Learn More
Medchemexpress LLC TGF beta 1/TGFB1 Hu 1mg
SDP
Catalog No. 5000196452
Encompass_Preferred
Recombinant human transforming growth factor beta 1 (TGF-β1) produced in CHO cells and supplied as a lyophilized powder. This multifunctional cytokine regulates cell growth, differentiation, and extracellular matrix production, and is intended for use in cell culture stimulation, signaling pathway analysis, and functional bioassays.
  • Produced in CHO cells for mammalian-like post-translational modifications.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • High biological activity (ED50 <0.2 ng/mL in standard cell inhibition assays).
  • Lyophilized formulation for extended shelf life and convenient storage.
  • Stable at -20 °C for long-term storage; short-term stability at 4 °C after reconstitution.
  • Suitable for cell culture stimulation, signaling studies, and bioactivity assays.
Catalog No. 50-001-96452 Supplier Medchemexpress LLC Supplier No. HYP71181MG
Recombinant human transforming growth factor beta 1 (TGF-β1) produced in CHO cells and supplied as a lyophilized powder. This multifunctional cytokine regulates cell growth, differentiation, and extracellular matrix production, and is intended for use in cell culture stimulation, signaling pathway analysis, and functional bioassays.
  • Produced in CHO cells for mammalian-like post-translational modifications.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • High biological activity (ED50 <0.2 ng/mL in standard cell inhibition assays).
  • Lyophilized formulation for extended shelf life and convenient storage.
  • Stable at -20 °C for long-term storage; short-term stability at 4 °C after reconstitution.
  • Suitable for cell culture stimulation, signaling studies, and bioactivity assays.