Learn More
Medchemexpress LLC TGF beta 1/TGFB1 Hu 20ug
SDP
Catalog No. 5000196548
Encompass_Preferred
Recombinant human Transforming Growth Factor-β1 (TGF-β1) is supplied as an N-Avi biotinylated protein expressed in mammalian cells and provided in a 20 μg vial for in-vitro research. The biotinylated Avi tag enables streptavidin-based capture or detection while mammalian expression supports native folding and post-translational modifications.
  • Recombinant human TGF-β1 with Avi biotin tag for streptavidin capture or detection.
  • Expressed in mammalian cells to support native post-translational modifications.
  • Provided in a 20 μg research-scale vial suitable for in-vitro assays.
  • Compatible with ELISA, receptor binding, and cell signaling assays.
  • Facilitates immobilization and sensitive detection in biochemical assays.
Catalog No. 50-001-96548 Supplier Medchemexpress LLC Supplier No. HYP7105620UG
Add to Cart
Add to Cart
Recombinant human Transforming Growth Factor-β1 (TGF-β1) is supplied as an N-Avi biotinylated protein expressed in mammalian cells and provided in a 20 μg vial for in-vitro research. The biotinylated Avi tag enables streptavidin-based capture or detection while mammalian expression supports native folding and post-translational modifications.
  • Recombinant human TGF-β1 with Avi biotin tag for streptavidin capture or detection.
  • Expressed in mammalian cells to support native post-translational modifications.
  • Provided in a 20 μg research-scale vial suitable for in-vitro assays.
  • Compatible with ELISA, receptor binding, and cell signaling assays.
  • Facilitates immobilization and sensitive detection in biochemical assays.