Learn More
Medchemexpress LLC TGF beta 1/TGFB1 Hu 250ug
SDP
Catalog No. 5000196520
Encompass_Preferred
Recombinant human transforming growth factor-beta 1 (TGF-β1) produced in CHO cells, supplied as a lyophilized, biologically active protein for research applications. It modulates cell proliferation, differentiation, migration, and immune responses, and is used in studies of development, fibrosis, and cancer research.
  • Produced in CHO cells for appropriate mammalian expression and post-translational modifications.
  • Lyophilized powder for convenient storage and transport.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive cell assays.
  • High biological potency with ED50 values in the sub-ng to pg/mL range and specific activity >1x10^7 units/mg.
  • Stable when stored frozen; recommended aliquoting for long-term use.
  • Lot-specific certificate of analysis available for buffer, concentration, and activity details.
Catalog No. 50-001-96520 Supplier Medchemexpress LLC Supplier No. HYP7118250UG
Recombinant human transforming growth factor-beta 1 (TGF-β1) produced in CHO cells, supplied as a lyophilized, biologically active protein for research applications. It modulates cell proliferation, differentiation, migration, and immune responses, and is used in studies of development, fibrosis, and cancer research.
  • Produced in CHO cells for appropriate mammalian expression and post-translational modifications.
  • Lyophilized powder for convenient storage and transport.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive cell assays.
  • High biological potency with ED50 values in the sub-ng to pg/mL range and specific activity >1x10^7 units/mg.
  • Stable when stored frozen; recommended aliquoting for long-term use.
  • Lot-specific certificate of analysis available for buffer, concentration, and activity details.