Learn More
Medchemexpress LLC TGF beta 1/TGFB1 LAP 10ug | 10ug
SDP
Catalog No. 5000194143
Encompass_Preferred
TGF beta 1 is a secreted ligand of the TGF-beta superfamily that binds to receptors and activates SMAD transcription factors Preproprotein is proteolytically processed to produce latency-associated peptide (LAP) and mature peptide TGF beta 1/TGFB1 LAP Protein Human (HEK293 N-His) is the recombinant human-derived TGF beta 1/TGFB1 Latency-associated peptide protein expressed by HEK293 with N-6 His labeled tag and C33S mutation
Catalog No. 50-001-94143 Supplier Medchemexpress LLC Supplier No. HYP7866810UG
TGF beta 1 is a secreted ligand of the TGF-beta superfamily that binds to receptors and activates SMAD transcription factors Preproprotein is proteolytically processed to produce latency-associated peptide (LAP) and mature peptide TGF beta 1/TGFB1 LAP Protein Human (HEK293 N-His) is the recombinant human-derived TGF beta 1/TGFB1 Latency-associated peptide protein expressed by HEK293 with N-6 His labeled tag and C33S mutation