Learn More
Medchemexpress LLC TGF beta 1/TGFB1 LAP 20ug
SDP
Catalog No. 5000180111
Encompass_Preferred
TGF beta 1/TGFB1 is a polypeptide member of the transforming growth factor beta superfamily of cytokines TGF beta 1 is a secreted protein that performs many cellular functions including the control of cell growth cell proliferation cell differentiation apoptosis and can regulate the expression and activation of other growth factors including interferon gamma and tumor necrosis factor alpha TGF beta 1/TGFB1 LAP Protein Human (HEK293 His) is the recombinant human-derived TGF beta 1/TGFB1 Latency-associated peptide protein expressed by HEK293 with C-10 His labeled tag and C33S mutation
Catalog No. 50-001-80111 Supplier Medchemexpress LLC Supplier No. HYP7361620UG
TGF beta 1/TGFB1 is a polypeptide member of the transforming growth factor beta superfamily of cytokines TGF beta 1 is a secreted protein that performs many cellular functions including the control of cell growth cell proliferation cell differentiation apoptosis and can regulate the expression and activation of other growth factors including interferon gamma and tumor necrosis factor alpha TGF beta 1/TGFB1 LAP Protein Human (HEK293 His) is the recombinant human-derived TGF beta 1/TGFB1 Latency-associated peptide protein expressed by HEK293 with C-10 His labeled tag and C33S mutation