Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Transforming growth factor beta 1 latency-associated peptide (human) | >95.0% | 2UG
SDP

Catalog No. 5000174252
Encompass_Preferred

Recombinant human transforming growth factor beta-1 (TGF-β1) latency-associated peptide (LAP). This tag-free protein contains a C33S mutation and corresponds to residues L30-R278 (249 amino acids). It is expressed in HEK293 cells, supplied in small vial sizes (2 μg), and tested at >95% purity by reducing SDS-PAGE. Predicted molecular mass is 28.5 kDa; SDS-PAGE typically shows a 20-45 kDa migration. Store frozen and handle per the supplier datasheet.

  • Tag-free latency-associated peptide with defined C33S mutation
  • Produced in HEK293 expression system for proper folding and post-translational modifications
  • High purity (>95%) suitable for biochemical and cell-based assays
  • Predicted molecular mass 28.5 kDa with expected SDS-PAGE profile of 20-45 kDa
  • Provided in small aliquots for low-volume experiments
  • Stable when aliquoted and stored at -20°C or -80°C

Catalog No. 50-001-74252 Supplier Medchemexpress LLC Supplier No. HYP704332UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human transforming growth factor beta-1 (TGF-β1) latency-associated peptide (LAP). This tag-free protein contains a C33S mutation and corresponds to residues L30-R278 (249 amino acids). It is expressed in HEK293 cells, supplied in small vial sizes (2 μg), and tested at >95% purity by reducing SDS-PAGE. Predicted molecular mass is 28.5 kDa; SDS-PAGE typically shows a 20-45 kDa migration. Store frozen and handle per the supplier datasheet.

  • Tag-free latency-associated peptide with defined C33S mutation
  • Produced in HEK293 expression system for proper folding and post-translational modifications
  • High purity (>95%) suitable for biochemical and cell-based assays
  • Predicted molecular mass 28.5 kDa with expected SDS-PAGE profile of 20-45 kDa
  • Provided in small aliquots for low-volume experiments
  • Stable when aliquoted and stored at -20°C or -80°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.