Learn More
Medchemexpress LLC TGF beta 1/TGFB1 Mo 100ug
SDP
Supplier:  Medchemexpress LLC HYP7117100UG
Encompass_Preferred
Recombinant mouse/rat transforming growth factor beta 1 (TGF-β1), expressed in HEK293 cells and supplied tag-free, for use in cell-based assays to study TGF-β signaling, cell cycle regulation, and apoptosis. Follow manufacturer instructions for reconstitution and storage.
  • Expressed in HEK293 cells for mammalian-like post-translational modifications.
  • Tag-free preparation for studies requiring native protein.
  • Demonstrated bioactivity: ED50 5-25 pg/mL (TF-1 assay) and <0.8 ng/mL (Mv-1-Lu assay).
  • Low endotoxin level (<1 EU/μg) suitable for sensitive applications.
  • Stable at -20°C; after reconstitution stable at 4°C for 1 week; aliquot and store at -20°C or -80°C for long-term storage.
Catalog No. 50-001-96467