Learn More
Medchemexpress LLC TGF beta 1/TGFB1 Mo 250ug
SDP
Catalog No. 5000196450
Encompass_Preferred
Recombinant transforming growth factor beta 1 (TGF-β1) derived from mouse/rat sequence, expressed in HEK293 cells and supplied as a tag-free, lyophilized protein. It is formulated for cell signaling and bioactivity assays, providing defined potency, low endotoxin, and high purity for reproducible in vitro experiments.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Tag-free, HEK293-expressed protein to support native folding and post-translational modifications.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive cell-based assays.
  • Defined biological activity (ED50 in the pg-ng/mL range) for potency benchmarking.
  • Lyophilized from 4 mM HCl for extended shelf life and convenient storage.
  • Recommended aliquoting and carrier protein addition for long-term stability.
  • Available in multiple pack sizes, including a 250 μg option.
Catalog No. 50-001-96450 Supplier Medchemexpress LLC Supplier No. HYP7117250UG
Recombinant transforming growth factor beta 1 (TGF-β1) derived from mouse/rat sequence, expressed in HEK293 cells and supplied as a tag-free, lyophilized protein. It is formulated for cell signaling and bioactivity assays, providing defined potency, low endotoxin, and high purity for reproducible in vitro experiments.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Tag-free, HEK293-expressed protein to support native folding and post-translational modifications.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive cell-based assays.
  • Defined biological activity (ED50 in the pg-ng/mL range) for potency benchmarking.
  • Lyophilized from 4 mM HCl for extended shelf life and convenient storage.
  • Recommended aliquoting and carrier protein addition for long-term stability.
  • Available in multiple pack sizes, including a 250 μg option.