Learn More
Medchemexpress LLC TGF beta 1/TGFB1 Mo 2ug

Supplier: Medchemexpress LLC HYP71172UG
Recombinant TGF-β1 protein (mouse/rat), expressed in HEK293 cells and supplied as a lyophilized, tag-free polypeptide corresponding to residues A279-S390. The protein is >95% pure by reducing SDS-PAGE, has an approximate molecular weight of 13 kDa, and demonstrates potent biological activity in standard cell proliferation assays (ED50 in the pg-ng/mL range). Formulated from a 0.22 μm filtered solution of 4 mM HCl and intended for in vitro signaling and functional assays; store frozen and avoid repeated freeze-thaw by aliquoting.
- Recombinant protein expressed in HEK293 cells.
- Tag-free, representing residues A279-S390.
- Purity greater than 95% by reducing SDS-PAGE.
- Approximate molecular weight ~13 kDa.
- Lyophilized formulation from 4 mM HCl.
- Potent biological activity: ED50 in pg-ng/mL range.
- Store frozen and aliquot to minimize freeze-thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.