Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TGF beta 1/TGFB1 Mo 500ug
SDP

Catalog No. 5000196615
Encompass_Preferred

Recombinant TGF-β1 (mature A279-S390), produced in HEK293 cells and supplied lyophilized. Designed for in vitro functional and cell-based assays that measure TGF-β activity, the protein is provided with validated biological activity and low endotoxin levels for reliable experimental results.

  • Recombinant mature TGF-β1 (A279-S390) for functional assays.
  • Expression in HEK293 cells preserves native post-translational modifications.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Low endotoxin (<1 EU/μg) suitable for cell-based studies.
  • Lyophilized formulation for long-term storage and transport.
  • Stability when aliquoted and stored at -20°C or -80°C for extended periods.
  • Validated biological activity with ED50 values reported in standard cell assays.

Catalog No. 50-001-96615 Supplier Medchemexpress LLC Supplier No. HYP7117500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant TGF-β1 (mature A279-S390), produced in HEK293 cells and supplied lyophilized. Designed for in vitro functional and cell-based assays that measure TGF-β activity, the protein is provided with validated biological activity and low endotoxin levels for reliable experimental results.

  • Recombinant mature TGF-β1 (A279-S390) for functional assays.
  • Expression in HEK293 cells preserves native post-translational modifications.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Low endotoxin (<1 EU/μg) suitable for cell-based studies.
  • Lyophilized formulation for long-term storage and transport.
  • Stability when aliquoted and stored at -20°C or -80°C for extended periods.
  • Validated biological activity with ED50 values reported in standard cell assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.