Learn More
Alpco Ltd TGF Beta-2 ELISA 10 L
SDP
Supplier:  Alpco Ltd 20TGF2HUE01
Encompass_Preferred
The TGF-?2 ELISA is an enzyme immunoassay for the quantitative measurement of TGF-?2 in serum, plasma, and cell culture supernatant. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3531