Learn More
Medchemexpress LLC TGF beta 2/TGFB2 Rh 100ug
SDP
Supplier:  Medchemexpress LLC HYP73613A100UG
Encompass_Preferred
Transforming growth factor-beta 2 (TGF- 2) an extracellular glycosylated protein is a member of the TGF- superfamily TGF 2 controls key physiological processes including cell migration proliferation and differentiation via signalling through type I and type II receptors (TGF R1 and TGF R2) TGF- 2 is an immune suppressor involved in the development of immune tolerance and also regulates embryonic development[1][2] TGF beta 2/TGFB2 Protein Rhesus Macaque (HEK293 His) is produced in HEK293 cells with a C-Terminal His-tag It and consists of 414 amino acids (M1-S414)
Catalog No. 50-001-79972