Learn More
Lonza Walkersville TGF Beta 3 for hMSC Chondrogenic Differentiation Kit
Catalog No. NC9996853
Encompass
TGF Beta 3 required for use with the hMSC Chondrogenic Differentiation BulletKit (PT-3003).Content:1 x Lyophilized TGF Beta 3, 2 µg
Catalog No. NC9996853 Supplier Lonza Walkersville Supplier No. PT4124
TGF Beta 3 required for use with the hMSC Chondrogenic Differentiation BulletKit (PT-3003).Content:1 x Lyophilized TGF Beta 3, 2 µg