Learn More
Abcam Recombinant human TGF beta 1 protein (Active), 10UG
Supplier:  Abcam AB5003610UG
Encompass_Preferred
Recombinant human TGF beta 1 protein (Active) is a Human Full Length TGF-beta 1 protein, in the 279 to 390 aa range with >98% purity, < 1 EU/µg endotoxin level and suitable for Cell Culture, Functional studies, western blot, HPLC and SDS-PAGE. The predicted molecular weight of ab50036 recombinant protein is 25 kDa. - Save time and ensure accurate results - using our recombinant TGF-beta 1 cytokine protein - Optimal protein bioactivity, stability and reproducibility - Available in different sizes to fit your experimental needs - Cited in over 60 publications
Protein/ Peptide/ Enzyme Synonyms: TGFB, TGFB1, Transforming growth factor beta-1 proprotein
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0983488