Learn More
Santa Cruz Biotechnology TGF beta Receptor 2/TGFBR2 Antibody (E-6) 200 µg/ml
Catalog No. NC3785877
Encompass
TGF beta Receptor 2/TGFBR2 Antibody (E-6) is a mouse monoclonal IgG2a κ, cited in 68 publications, provided at 200 µg/mlraised against amino acids 1-567 representing full length TFGβ RII of human originAnti-TGF beta Receptor 2/TGFBR2 Antibody (E-6) is recommended for detection of TGFβ RII of mouse, rat and human origin by WB, IP, IF, IHC(P) and ELISA
Catalog No. NC3785877 Supplier Santa Cruz Biotechnology Supplier No. SC17792
TGF beta Receptor 2/TGFBR2 Antibody (E-6) is a mouse monoclonal IgG2a κ, cited in 68 publications, provided at 200 µg/mlraised against amino acids 1-567 representing full length TFGβ RII of human originAnti-TGF beta Receptor 2/TGFBR2 Antibody (E-6) is recommended for detection of TGFβ RII of mouse, rat and human origin by WB, IP, IF, IHC(P) and ELISA