Learn More
Medchemexpress LLC HY-P7428 10ug Medchemexpress, TGF-beta receptor 2, His (HEK293-expressed), Mouse CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP742810UG
Encompass_Preferred
Medchemexpress, HY-P7428 10ug TGF-beta receptor 2, His (HEK293-expressed), Mouse CAS: Recombinant Mouse Transforming growth factor-beta receptor type 2, His consists of N-terminal ectodomain and C-terminal protein kinase domain with his tag. TGF-beta receptor type-2 is the ligand-binding receptor for all members of the TGF-β family. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3163