Learn More
ABclonal Technology TGF beta Receptor I TGFBR1
SDP
Catalog No. 5000007336
Encompass_Preferred
The protein encoded by this gene forms a heteromeric complex with type II TGF-beta receptors when bound to TGF-beta transducing the TGF-beta signal from the cell surface to the cytoplasm The encoded protein is a serine threonine protein kinase Mutations in this gene have been associated with Loeys-Dietz aortic aneurysm syndrome (LDAS) Multiple transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-07336 Supplier ABclonal Technology Supplier No. A22151100UL
The protein encoded by this gene forms a heteromeric complex with type II TGF-beta receptors when bound to TGF-beta transducing the TGF-beta signal from the cell surface to the cytoplasm The encoded protein is a serine threonine protein kinase Mutations in this gene have been associated with Loeys-Dietz aortic aneurysm syndrome (LDAS) Multiple transcript variants encoding different isoforms have been found for this gene