Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TGF beta Receptor II Antibody | 100 UL
SDP

Catalog No. 5000442423
Encompass_Preferred

This TGF beta Receptor II Antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody that targets TGF beta Receptor II.

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Applications: WB, ELISA, IHC-P, ICC/IF
  • Clonality: Polyclonal
  • Isotype: IgG
  • Molecular weight: 65 kDa
  • Purity: Affinity purified
  • Formulation: Supplied in PBS containing 50% glycerol, 0.5% BSA and 0.02% sodium azide, pH 7.3
  • Storage: Stored at -20°C for 1 year. Avoid repeated freeze / thaw cycles.

Catalog No. 50-004-42423 Supplier Medchemexpress LLC Supplier No. HYP81131100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This TGF beta Receptor II Antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody that targets TGF beta Receptor II.

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Applications: WB, ELISA, IHC-P, ICC/IF
  • Clonality: Polyclonal
  • Isotype: IgG
  • Molecular weight: 65 kDa
  • Purity: Affinity purified
  • Formulation: Supplied in PBS containing 50% glycerol, 0.5% BSA and 0.02% sodium azide, pH 7.3
  • Storage: Stored at -20°C for 1 year. Avoid repeated freeze / thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.