Learn More
Biotium TGF-beta (Transforming Growth Factor beta)(TGFB/510), CF740 conjugate, 0.1mg/mL
Catalog No. 5000591224
Encompass_Preferred
This MAb recognizes TGF beta 1, 2 and 3. TGFs inhibit the growth of epithelial cells and stimulate the growth of mesenchymal cells. Most breast lesions, benign and malignant, involve abnormal proliferation and altered architecture of stromal and/or epithelial elements.
Catalog No. 50-005-91224 Supplier Biotium Supplier No. BNC740510500
Add to Cart
Add to Cart
This MAb recognizes TGF beta 1, 2 and 3. TGFs inhibit the growth of epithelial cells and stimulate the growth of mesenchymal cells. Most breast lesions, benign and malignant, involve abnormal proliferation and altered architecture of stromal and/or epithelial elements.