Learn More
Biotium TGF-beta (Transforming Growth Factor beta)(TGFB/510), CF740 conjugate, 0.1mg/mL
Supplier: Biotium BNC740510100
This MAb recognizes TGF beta 1, 2 and 3. TGFs inhibit the growth of epithelial cells and stimulate the growth of mesenchymal cells. Most breast lesions, benign and malignant, involve abnormal proliferation and altered architecture of stromal and/or epithelial elements.