Learn More
Medchemexpress LLC HY-P7117 10ug Medchemexpress, TGF-beta1 (HEK293-expressed), Mouse CAS: Purity:>98%
SDP
Catalog No. 501934345
Encompass_Preferred
Medchemexpress, HY-P7117 10ug TGF-beta1 (HEK293-expressed), Mouse CAS: Recombinant Mouse TGF-beta 1/TGFB1 (HEK293-expressed) is a multifunctional cytokine that controls numerous biological processes including immunity, differentiation, tumor suppression, tumor metastasis, senescence, migration, wound healing, apoptosis, cell division, adipogenesis, and osteogenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4345 Supplier Medchemexpress LLC Supplier No. HYP711710UG
Medchemexpress, HY-P7117 10ug TGF-beta1 (HEK293-expressed), Mouse CAS: Recombinant Mouse TGF-beta 1/TGFB1 (HEK293-expressed) is a multifunctional cytokine that controls numerous biological processes including immunity, differentiation, tumor suppression, tumor metastasis, senescence, migration, wound healing, apoptosis, cell division, adipogenesis, and osteogenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.