Learn More
Enzo Life Sciences TGF-β1 (human), (recombinant) (20µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2210020
Encompass_Preferred
Alternative name: Differentiation inhibiting factor, Cartilage-inducing factor, Transforming growth factor beta1. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains 0.1% trifluoroacetic acid and trehalose (in a 20:1 trehalose to protein ratio). Source: Produced in CHO cells. Contains 112/224 amino acids. Long Term Storage: -80°C.
Catalog No. 50-201-4018