Learn More
ACROBiosystems ACROBiosystems ActiveMax Human TGF-Beta 1 / TGFB1 Protein, Tag Free
Supplier:  ACROBiosystems TG1H42121MG
Encompass_Preferred
Recombinant Protein;1MG;ActiveMax Human TGFB1, Tag Free (TG1-H4212) is expressed from human 293 cells (HEK293). It contains AA Ala 279 - Ser 390 (Accession # P01137-1).;This protein carries no "tag". The protein has a calculated MW of 12.8 kDa (monomer). the protein migrates as 14 kDa (monomer) under reducing (R) condition, and 25 kDa (Dimer) under non-reducing (NR) condition (SDS-PAGE).;Immobilized ActiveMax Human TGFB1, Tag Free (Cat. No. TG1-H4212) at 2 μg/mL (100 μL/well) can bind Human TGFBR2, Fc Tag (Cat. No. TG2-H5252) with a linear range of 0.6-5 ng/mL (QC tested).;ActiveMax Human TGFB1, Tag Free (Cat. No. TG1-H4212) inhibits the IL-4-dependent proliferation of TF-1 cells. The ED50 for this effect is 0.390-0.476 pg/ml (Routinely tested).
Catalog No. 50-195-7553