Learn More
Medchemexpress LLC HY-P7119 10ug Medchemexpress, TGF-beta2, Human CAS: Purity:>98%
SDP
Catalog No. 501934347
Encompass_Preferred
Medchemexpress, HY-P7119 10ug TGF-beta2, Human CAS: Recombinant Human Transforming Growth Factor β2 is a multifunctional cytokine which participates in control of cell growth and differentiation, induces fibrosis, causes the suppression of immune response and has a vital role during embryonic development. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4347 Supplier Medchemexpress LLC Supplier No. HYP711910UG
Medchemexpress, HY-P7119 10ug TGF-beta2, Human CAS: Recombinant Human Transforming Growth Factor β2 is a multifunctional cytokine which participates in control of cell growth and differentiation, induces fibrosis, causes the suppression of immune response and has a vital role during embryonic development. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.