Learn More
Enzo Life Sciences TGF-beta3 (mouse), (recombinant) (10µg)
SDP
Catalog No. 502008021
Encompass_Preferred
Alternative name: Transforming growth factor beta. Concentration: 0.25mg/ml. Endotoxin: ≤1 EU/μg protein measured by kinetic LAL analysis. Purity: ≥98% (Reducing and Non-reducing SDS-PAGE). MW: ~25.5kDa. Appearance: Clear solution. Formulation: Liquid. In 20% ethanol and 0.12% acetic acid. Source: Produced in E. coli. Non-glycosylated, disulfide-linked homodimer, containing two 112 amino acid chains. Long Term Storage: +4°C. Use/Stability: Stable at +4°C for 1 year from date of purchase.
Catalog No. 50-200-8021 Supplier Enzo Life Sciences Supplier No. ALX2018170010
Alternative name: Transforming growth factor beta. Concentration: 0.25mg/ml. Endotoxin: ≤1 EU/μg protein measured by kinetic LAL analysis. Purity: ≥98% (Reducing and Non-reducing SDS-PAGE). MW: ~25.5kDa. Appearance: Clear solution. Formulation: Liquid. In 20% ethanol and 0.12% acetic acid. Source: Produced in E. coli. Non-glycosylated, disulfide-linked homodimer, containing two 112 amino acid chains. Long Term Storage: +4°C. Use/Stability: Stable at +4°C for 1 year from date of purchase.