Learn More
Medchemexpress LLC TGF-βRI inhibitor 3 (compound 9ac) | 666729-57-3 | 99.2% | 351.40 g·mol⁻¹ | C21H21NO4 | 5 MG
SDP
Catalog No. 5000222088
Encompass_Preferred
TGF-βRI inhibitor 3 is a selective small-molecule inhibitor of the Transforming Growth Factor-β type I receptor used in research to study TGF-β signaling and related cellular processes. The compound is supplied as a light yellow solid with high purity and defined storage recommendations.
  • Selective inhibition of TGF-βRI signaling.
  • High purity (99.16%).
  • Chemical formula C21H21NO4 and molecular weight 351.40 g·mol⁻¹.
  • Provided as a light yellow solid suitable for laboratory use.
  • Available in milligram quantities for small-scale experiments.
  • Powder stable at -20°C for long-term storage; in solvent store at -80°C for extended periods.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-22088 Supplier Medchemexpress LLC Supplier No. HY1379185MG
TGF-βRI inhibitor 3 is a selective small-molecule inhibitor of the Transforming Growth Factor-β type I receptor used in research to study TGF-β signaling and related cellular processes. The compound is supplied as a light yellow solid with high purity and defined storage recommendations.
  • Selective inhibition of TGF-βRI signaling.
  • High purity (99.16%).
  • Chemical formula C21H21NO4 and molecular weight 351.40 g·mol⁻¹.
  • Provided as a light yellow solid suitable for laboratory use.
  • Available in milligram quantities for small-scale experiments.
  • Powder stable at -20°C for long-term storage; in solvent store at -80°C for extended periods.
  • For research use only; not for human or clinical use.