Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7039MSM2XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM542. Clonality: Monoclonal. Host: Mouse. Isotype: IgM kappa. Positive Control: Jurkat or Ramos cells, Heart, Kidney, Pituitary, Breast cancer, Melanoma. Cellular Staining: Cytoplasmic, Extracellular (secreted). Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: A 10-amino acid synthetic peptide (aa34-43; PPVAAAVVSH) from human TGFalpha-. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: ~6kDa.Chromosome: 2p13.3. Gene Symbol: TGFA. Gene ID: 7039. Ensemble: ENSG00000163235. Uniprot: NP_001295087.
Catalog No. 50-188-5211