Learn More
Aviva Systems Biology Corp TGFB1 Antibody - middle region (ARP37894_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37894_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TGFB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse TGFB1. Alternate names: Tgfb, Tgfb-1, TGFbeta1, TGF-beta1ApplicationIHC, WB, IF. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Mouse, Pig, Rat, Sheep
Catalog No. 50-014-56459