Learn More
Genscript Corporation TGFβ1, Bovine, 10ug, Purity: ≥ 95%
Catalog No. NC3787609
Encompass
TGF-β1 (transforming growth factor beta 1) is one of three closely related mammalian members of the large TGF-β1 superfamily that share a characteristic cystine knot structure. TGF-β1, -2 and -3 are highly pleiotropic
Catalog No. NC3787609 Supplier Genscript Corporation Supplier No. Z0377910
TGF-β1 (transforming growth factor beta 1) is one of three closely related mammalian members of the large TGF-β1 superfamily that share a characteristic cystine knot structure. TGF-β1, -2 and -3 are highly pleiotropic