Learn More
Aviva Systems Biology Corp TGFB3 Antibody - middle region (ARP45450_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45450_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TGFB3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TGFB3. Alternate names: ARVD, FLJ16571, TGF-beta3ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-59032