Learn More
Aviva Systems Biology Corp Tgfb3 Antibody - middle region (ARP45448_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45448_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tgfb3. It was validated on Western Blot. . Alternate names: MGC105479ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-285-201