Learn More
Sino Biological Recombinant Anti-TGF beta 1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10804R016100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human TGF-beta 1 (rh TGF-beta 1; Catalog#10804-H08H; NP_000651.3; Met 1-Ser 390).
Catalog No. 50-228-2512