Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human Latent TGF-Beta 1 / TGFB1 Protein, His Tag, Avi Tag (Avitag™);HEK293;200UG;TG1,TGFB1,TGF B1,TGF-B1,TGF-beta 1,TGF beta1,TGFbeta1,CED,DPD1,DPD-1,LAP,TGFB,TGFbeta,TGF-β1,TGFβ1,TGF-beta,TGF beta
Catalog No. 501657476
Encompass_Preferred
Recombinant Protein;Biotinylated Human Latent TGF-Beta 1 / TGFB1 Protein, His Tag, Avi Tag (Avitag™);200UG;Biotinylated Human Latent TGF-Beta 1, His Tag, Avi Tag (TG1-H82Qb) is expressed from human 293 cells (HEK293). It contains AA Leu 30 - Ser 390 (Accession # P01137-1 (C33S)).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 45.0 kDa. The protein migrates as 14 kDa, 40-48 kDa and 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ITGAV&ITGB6, His Tag (Cat. No. IT6-H52E1) at 5 μg/mL (100 μL/well) can bind Biotinylated Human Latent TGF-Beta 1, His Tag, Avi Tag (Cat. No. TG1-H82Qb) with a linear range of 0.5-4 ng/mL (QC tested).;Immobilized Human ITGAV&ITGB8, His Tag (Cat. No. IT8-H52W4) at 5 μg/mL (100 μL/well) can bind Biotinylated Human Latent TGF-Beta 1, His Tag, Avi Tag (Cat. No. TG1-H82Qb) with a linear range of 0.2-4 ng/mL (Routinely tested).
Catalog No. 50-165-7476 Supplier ACROBiosystems Supplier No. TG1H82QB200UG
Recombinant Protein;Biotinylated Human Latent TGF-Beta 1 / TGFB1 Protein, His Tag, Avi Tag (Avitag™);200UG;Biotinylated Human Latent TGF-Beta 1, His Tag, Avi Tag (TG1-H82Qb) is expressed from human 293 cells (HEK293). It contains AA Leu 30 - Ser 390 (Accession # P01137-1 (C33S)).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 45.0 kDa. The protein migrates as 14 kDa, 40-48 kDa and 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ITGAV&ITGB6, His Tag (Cat. No. IT6-H52E1) at 5 μg/mL (100 μL/well) can bind Biotinylated Human Latent TGF-Beta 1, His Tag, Avi Tag (Cat. No. TG1-H82Qb) with a linear range of 0.5-4 ng/mL (QC tested).;Immobilized Human ITGAV&ITGB8, His Tag (Cat. No. IT8-H52W4) at 5 μg/mL (100 μL/well) can bind Biotinylated Human Latent TGF-Beta 1, His Tag, Avi Tag (Cat. No. TG1-H82Qb) with a linear range of 0.2-4 ng/mL (Routinely tested).