Learn More
Santa Cruz Biotechnology TGFbeta3 Antibody (G-9), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC166833
Encompass
Transforming growth factor betas (TGFbetas) were originally discovered due to their ability to promote anchorage-independent growth of rat NRK fibroblasts in the presence of TGFbeta. TGFbeta1, TGFbeta2 and TGFbeta3 are each synthesized as precursor proteins that are very similar in that each is cleaved to yield a 112 amino acid polypeptide that remains associated with the latent portion of the molecules.
Catalog No. NC1764842