Learn More
ACROBiosystems Recombinant Protein;Human TGF-beta RII / TGFBR2 Protein, Fc Tag;HEK293;100UG;TG2,TGFBR2,TGFR2,TGFBR-2,TGFR-2,TbetaR-II,TGFβR2,TGFβ R2,TGFB R2,TGF-beta R2
Catalog No. 501658615
Encompass_Preferred
Recombinant Protein;Human TGF-beta RII / TGFBR2 Protein, Fc Tag;100UG;Human TGFBR2, Fc Tag (TG2-H5252) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Asp 159 (Accession # NP_003233).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.0 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8615 Supplier ACROBiosystems Supplier No. TG2H5252100UG
Recombinant Protein;Human TGF-beta RII / TGFBR2 Protein, Fc Tag;100UG;Human TGFBR2, Fc Tag (TG2-H5252) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Asp 159 (Accession # NP_003233).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.0 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.