Learn More
Aviva Systems Biology Corp TGFBI Antibody - C-terminal region (ARP44268_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44268_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TGFBI. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TGFBI. Alternate names: BIGH3, CDB1, CDG2, CDGG1, CSD, CSD1, CSD2, CSD3, EBMD, LCD1ApplicationIF, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58763