Learn More
Medchemexpress LLC TGFBI protein, human (HEK293, His) | >80.0% | 10 UG
SDP
Catalog No. 5000293535
Encompass_Preferred
Recombinant human transforming growth factor-beta-induced protein (TGFBI) produced in HEK293 cells and supplied as a solution with a C-terminal His tag. Intended for research use in studies of cell adhesion, extracellular matrix interactions, and biochemical characterization. Purity is stated as greater than 80% by reducing SDS-PAGE.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • C-terminal His tag enables purification and detection.
  • Supplied as a solution ready for downstream assays.
  • Purity >80% as determined by reducing SDS-PAGE.
  • Suitable for studies of cell adhesion and extracellular matrix interactions.
Catalog No. 50-002-93535 Supplier Medchemexpress LLC Supplier No. HYP7452510UG
Recombinant human transforming growth factor-beta-induced protein (TGFBI) produced in HEK293 cells and supplied as a solution with a C-terminal His tag. Intended for research use in studies of cell adhesion, extracellular matrix interactions, and biochemical characterization. Purity is stated as greater than 80% by reducing SDS-PAGE.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • C-terminal His tag enables purification and detection.
  • Supplied as a solution ready for downstream assays.
  • Purity >80% as determined by reducing SDS-PAGE.
  • Suitable for studies of cell adhesion and extracellular matrix interactions.