Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TGFBR1 (ALK5), GST Tag Recombinant Protein

Catalog No. PR9075A
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR9075A 100 μg
PV5837 10 μg
PV5838 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PR9075A Supplier Invitrogen™ Supplier No. PR9075A
Add to Cart
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

TGFBR1 is a transmembrane serine/threonine kinase in the TGFBR1 superfamily, which also includes ACVRs and BMPRs. Upon binding TGF-b, TGFBR2 dimerizes with and activates TGFBR1 via phosphorylation leading to downstream phosphorylation of SMADs by TGFBR1. TGFBR1 forms a heteromeric complex with type II TGF-beta receptors when bound to TGF-beta, transducing the TGF-beta signal from the cell surface to the cytoplasm. The encoded protein is a serine/threonine protein kinase. Mutations in this gene have been associated with Loeys-Dietz aortic aneurysm syndrome (LDAS).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P36897
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with no preservative
Gene ID (Entrez) 7046
Molecular Weight (g/mol) 62.9 kDa
Name Human TGFBR1 (ALK5), GST Tag
Purification Method Purified
Quantity 100 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AAT5; activin A receptor type II-like kinase, 53kDa; activin A receptor type II-like protein kinase of 53kD; Activin receptor-like kinase 5; ACVRLK4; ALK5; Alk-5; AU017191; ESK2; ESS1; LDS1; LDS1A; LDS2A; MSSE; mutant transforming growth factor beta receptor I; ser; serine/threonine-protein kinase receptor R4; SKR4; TbetaRI; tbetaR-I; tgf beta receptor 1; TGF-beta receptor type I; TGF-beta receptor type-1; TGF-beta type I receptor; TGFBR1; TGFR1; TGFR-1; Transfor; transforming growth factor beta receptor 1; transforming growth factor beta receptor I; transforming growth factor, beta receptor 1; transforming growth factor, beta receptor I; transforming growth factor, beta receptor I (activin A receptor type II-like kinase, 53kDa); transforming growth factor-beta receptor type I
Common Name TGFBR1
Gene Symbol TGFBR1
Kinase Family STKR Kinase Family
Protein Length 1-503
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 1-503
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Ser⁄Thr Kinases: TKL Group
Shipping Condition Dry Ice
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.