Learn More
Aviva Systems Biology Corp TGFBR2 Antibody - N-terminal region (ARP44743_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44743_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TGFBR2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TGFBR2. Alternate names: HNPCC6, MFS2, RIIC, Tbeta, AAT3, FAA3, LDS1B, LDS2B, TAAD2, TGFR-2, TGFbeta-RIIApplicationWB. Species reactivity: Cow, Goat, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-58889