Learn More
Sino Biological Recombinant Human TGF-beta RII/TGFBR2 Protein (ECD, His & hFc Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10358H03HB20
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Asp159) of human TGFB receptor 2 (NP_003233.4) was expressed with the fused C-terminal His-tagged Fc region of human IgG1 at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-5697