Learn More
Invitrogen™ Human TGFBR2, GST Tag Recombinant Protein
Description
Specifications
Specifications
| Accession Number | P37173 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM tris HCl with 150mM NaCl, 0.5mM EDTA, 2mM DTT, 50% glycerol, 0.02% NP-40 and no preservative; pH 7.5 |
| Gene ID (Entrez) | 7048 |
| Molecular Weight (g/mol) | 70.3 kDa |
| Name | Human TGFBR2, GST Tag |
| pH Range | 7.5 |
| Purification Method | Purified |
| Quantity | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.