Learn More
Medchemexpress LLC TGFBR2/TGF-beta RII 10ug
SDP
Catalog No. 5000180657
Encompass_Preferred
TGFBR2 is a transmembrane kinase that forms the TGF- receptor complex with TGFBR1 and regulates various processes Upon binding to cytokines this complex activates TGFBR2 initiating SMAD-dependent signaling through phosphorylation of TGFBR1 SMAD2 and SMAD4 TGFBR2/TGF-beta RII Protein Rat (HEK293 Fc) is the recombinant rat-derived TGFBR2/TGF-beta RII protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-80657 Supplier Medchemexpress LLC Supplier No. HYP7343010UG
TGFBR2 is a transmembrane kinase that forms the TGF- receptor complex with TGFBR1 and regulates various processes Upon binding to cytokines this complex activates TGFBR2 initiating SMAD-dependent signaling through phosphorylation of TGFBR1 SMAD2 and SMAD4 TGFBR2/TGF-beta RII Protein Rat (HEK293 Fc) is the recombinant rat-derived TGFBR2/TGF-beta RII protein expressed by HEK293 with C-hFc labeled tag