Learn More
Medchemexpress LLC TGFBR2/TGF-beta RII 2ug
SDP
Catalog No. 5000180658
Encompass_Preferred
The TGFBR2/TGF- RII protein cooperates with TGFBR1 to form dedicated receptors for TGFB1 TGFB2 and TGFB3 It serves as a signal transducer that coordinates diverse physiological and pathological processes including cell cycle regulation mesenchymal cell dynamics wound healing matrix production immunosuppression and carcinogenesis TGFBR2/TGF-beta RII Protein Human (sf9 His) is the recombinant human-derived TGFBR2/TGF-beta RII protein expressed by Sf9 insect cells with C-His labeled tag
Catalog No. 50-001-80658 Supplier Medchemexpress LLC Supplier No. HYP734282UG
The TGFBR2/TGF- RII protein cooperates with TGFBR1 to form dedicated receptors for TGFB1 TGFB2 and TGFB3 It serves as a signal transducer that coordinates diverse physiological and pathological processes including cell cycle regulation mesenchymal cell dynamics wound healing matrix production immunosuppression and carcinogenesis TGFBR2/TGF-beta RII Protein Human (sf9 His) is the recombinant human-derived TGFBR2/TGF-beta RII protein expressed by Sf9 insect cells with C-His labeled tag