Learn More
Medchemexpress LLC TGFBR2/TGF-beta RII 500ug
SDP
Catalog No. 5000178608
Encompass_Preferred
TGF-beta receptor 2 Protein Human (HEK293 Fc) a fragment of transforming growth factor-beta receptor type 2 (T RII) consists of amino acid of Thr23-Asp159 with an Fc tag at the C-terminus
Catalog No. 50-001-78608 Supplier Medchemexpress LLC Supplier No. HYP7426500UG
TGF-beta receptor 2 Protein Human (HEK293 Fc) a fragment of transforming growth factor-beta receptor type 2 (T RII) consists of amino acid of Thr23-Asp159 with an Fc tag at the C-terminus