Learn More
Carnabio Usa Inc TGFBR2(TGFBR2)/10UG/CATALYTIC
Supplier:  Carnabio Usa Inc 0914210
Encompass
TGFR2(TGFBR2), Catalytic domain, Wild type, Amino Acid 194-567 , NP_003233.2, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-1004