Learn More
Medchemexpress LLC TGFBR3/TGF-beta RIII 50ug
SDP
Catalog No. 5000183044
Encompass_Preferred
TGFBR3/TGF-beta RIII Protein is pivotal binding to TGF-beta indicating a potential role in capturing and presenting TGF-beta to signaling receptors In gonadotrope cells it acts as an inhibin A coreceptor regulating FSH levels and impacting female fertility Its interaction with DYNLT4 emphasizes diverse functional associations in cellular processes suggesting a multifaceted role for TGFBR3 in cellular regulation and fertility control TGFBR3/TGF-beta RIII Protein Mouse (HEK293 His) is the recombinant mouse-derived TGFBR3/TGF-beta RIII protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-83044 Supplier Medchemexpress LLC Supplier No. HYP7245250UG
TGFBR3/TGF-beta RIII Protein is pivotal binding to TGF-beta indicating a potential role in capturing and presenting TGF-beta to signaling receptors In gonadotrope cells it acts as an inhibin A coreceptor regulating FSH levels and impacting female fertility Its interaction with DYNLT4 emphasizes diverse functional associations in cellular processes suggesting a multifaceted role for TGFBR3 in cellular regulation and fertility control TGFBR3/TGF-beta RIII Protein Mouse (HEK293 His) is the recombinant mouse-derived TGFBR3/TGF-beta RIII protein expressed by HEK293 with C-6 His labeled tag