Learn More
Aviva Systems Biology Corp TGIF1 Antibody - C-terminal region (ARP51343_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51343_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TGIF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TGIF1. Alternate names: HPE4, MGC39747, MGC5066, TGIFApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-60554