Learn More
Aviva Systems Biology Corp TGIF2LY Antibody - middle region (ARP36141_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36141_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TGIF2LY. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TGIF2LY. Alternate names: TGIFLYApplicationWB. Species reactivity: Human, Yeast
Catalog No. 50-014-56028