Learn More
Aviva Systems Biology Corp TGIFX1 Antibody - middle region (ARP37505_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37505_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TGIFX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse TGIFX1. Alternate names: Tex1, Tgifx1, Tgif2lx, OTTMUSG00000018465ApplicationWB. Species reactivity: Mouse
Catalog No. 50-014-53077