Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7052MSM1XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM592. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: HUVEC Cells. Endothelial cells in placenta, liver, brain, or breast carcinoma. Smooth muscle cells of any origin (e.g. intestine). Cellular Staining: Cytoplasmic, cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human TGM2 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 77-85kDa.Chromosome: 20q12. Gene Symbol: TGM2. Gene ID: 7052. Ensemble: ENSG00000198959. Uniprot: NP_004604.
Catalog No. 50-188-5217